accessibility menu, dialog, popup

UserName

CAS RN 212757-09-0

OO(R)(R)OBr

CAS RN 212757-09-0

IUPAC Name: (1R,2R)-2-(4-bromobenzoyl)cyclohexane-1-carboxylate
Synonyms: (1R,2R)-2-(4-bromobenzoyl)cyclohexane-1-carboxylate
Molecular Weight (g/mol): 310.17
Molecular Formula: C14H14BrO3
InChi Key: OVZXXISOLDHARA-VXGBXAGGSA-M
SMILES: [O-]C(=O)[C@@H]1CCCC[C@H]1C(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)