CAS RN 212755-76-5
IUPAC Name:
methyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Synonyms:
methyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate
Molecular Weight (g/mol):
246.19
Molecular Formula:
C11H9F3O3
InChi Key:
NMJOILIYLOHXGP-UHFFFAOYSA-N
SMILES:
COC(=O)CC(=O)C1=CC=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
Methyl 4-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™
Methyl 4-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™
CAS: 212755-76-5 Molecular Formula: C11H9F3O3 Molecular Weight (g/mol): 246.19 MDL Number: MFCD00216521 InChI Key: NMJOILIYLOHXGP-UHFFFAOYSA-N Synonym: methyl 4-trifluoromethyl benzoylacetate,methyl 4-trifluoromethylbenzoylacetate,methyl 3-oxo-3-4-trifluoromethyl phenyl propanoate,3-oxo-3-4-trifluoromethylphenyl propionic acid methyl ester,methyl 3-4-trifluoromethyl phenyl-3-oxopropanoate,pubchem2696,buttpark 18\10-41,methyl 4-trifluoromethyl benzoyl acetate PubChem CID: 735882 IUPAC Name: methyl 3-oxo-3-[4-(trifluoromethyl)phenyl]propanoate SMILES: COC(=O)CC(=O)C1=CC=C(C=C1)C(F)(F)F