CAS RN 210832-85-2
IUPAC Name:
2-bromo-1-[4-(morpholin-4-yl)phenyl]ethan-1-one
Synonyms:
2-bromo-1-[4-(morpholin-4-yl)phenyl]ethanone
Molecular Weight (g/mol):
284.15
Molecular Formula:
C12H14BrNO2
InChi Key:
OUGMZFJPRSTGMJ-UHFFFAOYSA-N
SMILES:
BrCC(=O)C1=CC=C(C=C1)N1CCOCC1
1
–
3
of
3
results
2
2-Bromo-1-(4-morpholinophenyl)-1-ethanone, 97%, Thermo Scientific™
CAS: 210832-85-2 Molecular Formula: C12H14BrNO2 Molecular Weight (g/mol): 284.15 MDL Number: MFCD03783555 InChI Key: OUGMZFJPRSTGMJ-UHFFFAOYSA-N Synonym: 2-bromo-1-4-morpholinophenyl-1-ethanone,2-bromo-1-4-morpholinophenyl ethanone,2-bromo-1-4-morpholin-4-ylphenyl ethanone,2-bromo-1-4-4-morpholinyl phenyl ethanone,2-bromo-1-4-morpholin-4-yl-phenyl ethanone,2-bromo-1-4-morpholin-4-yl phenyl ethanone,2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one,pubchem23338,acmc-1cqxm,alpha-bromo-4'-morpholinoacetophenone PubChem CID: 2795357 SMILES: BrCC(=O)C1=CC=C(C=C1)N1CCOCC1