accessibility menu, dialog, popup

UserName

CH3H3CCH3OONCH3H2N

CAS RN 21075-83-2

IUPAC Name: N-methyl(tert-butoxy)carbohydrazide
Synonyms: N-methyltert-butoxycarbohydrazide
Molecular Weight (g/mol): 146.19
Molecular Formula: C6H14N2O2
InChi Key: IHMQNZFRFVYNDS-UHFFFAOYSA-N
SMILES: CN(N)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results