CAS RN 21064-69-7
IUPAC Name:
4-nitrophenyl 2-(trimethylazaniumyl)ethyl phosphate
Synonyms:
4-nitrophenyl 2-(trimethylammonio)ethyl phosphate
Molecular Weight (g/mol):
304.24
Molecular Formula:
C11H17N2O6P
InChi Key:
NAIXASFEPQPICN-UHFFFAOYSA-N
SMILES:
C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-Nitrophenylphosphorylcholine
CAS: 21064-69-7 Molecular Formula: C11H17N2O6P Molecular Weight (g/mol): 304.24 MDL Number: MFCD00077834 InChI Key: NAIXASFEPQPICN-UHFFFAOYSA-N Synonym: p-nitrophenylphosphorylcholine,o-4-nitrophenylphosphoryl choline,4-nitrophenylphosphorylcholine,4-nitrophenyl 2-trimethylammonio ethyl phosphate,4-nitrophenylphorylcholine,o-4-nitrophenyl-phosphoryl choline,4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate,2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt PubChem CID: 4307994 IUPAC Name: (4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate SMILES: C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O