accessibility menu, dialog, popup

UserName

CH3OONCS

CAS RN 21055-37-8

IUPAC Name: methyl 2-isothiocyanatoacetate
Synonyms: methyl 2-isothiocyanatoacetate
Molecular Weight (g/mol): 131.15
Molecular Formula: C4H5NO2S
InChi Key: GOWGDPFDGIPFIK-UHFFFAOYSA-N
SMILES: COC(=O)CN=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)