accessibility menu, dialog, popup

UserName

OHCoOH

CAS RN 21041-93-0

IUPAC Name: λ²-cobalt(2+) dihydroxide
Synonyms: cobalt(II) hydroxide
Molecular Weight (g/mol): 92.95
Molecular Formula: CoH2O2
InChi Key: ASKVAEGIVYSGNY-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Co++]
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)