accessibility menu, dialog, popup

UserName

OSN

CAS RN 20949-81-9

IUPAC Name: 2-phenyl-1,3-thiazole-4-carbaldehyde
Synonyms: 2-phenyl-1,3-thiazole-4-carbaldehyde
Molecular Weight (g/mol): 189.23
Molecular Formula: C10H7NOS
InChi Key: OLLKCCGWRITPOV-UHFFFAOYSA-N
SMILES: O=CC1=CSC(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)