accessibility menu, dialog, popup

UserName

OH3COOH3COOH3COLu

CAS RN 207500-05-8

IUPAC Name: lutetium(3+) triacetate
Synonyms: lutetium(3+) triacetate
Molecular Weight (g/mol): 352.1
Molecular Formula: C6H9LuO6
InChi Key: YMPHWTSLMNFNFB-UHFFFAOYSA-K
SMILES: [Lu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)