accessibility menu, dialog, popup

UserName

ONONHN

CAS RN 2075-46-9

IUPAC Name: 4-nitro-1H-pyrazole
Synonyms: 4-nitropyrazole
Molecular Weight (g/mol): 113.08
Molecular Formula: C3H3N3O2
InChi Key: XORHNJQEWQGXCN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNN=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)