accessibility menu, dialog, popup

UserName

CAS RN 207398-95-6

IUPAC Name: disodium 2-[(carboxylatomethyl)amino]acetate hydrate
Synonyms: disodium hydrate iminodiacetate
Molecular Weight (g/mol): 195.08
Molecular Formula: C4H7NNa2O5
InChi Key: YSBGCHXLMBAKPV-UHFFFAOYSA-L
SMILES: O.[Na+].[Na+].[O-]C(=O)CNCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results