accessibility menu, dialog, popup

UserName

OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHCe

CAS RN 206996-61-4

IUPAC Name: cerium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: cerium acetylacetonate
Molecular Weight (g/mol): 437.44
Molecular Formula: C15H21CeO6
InChi Key: LCRQZRNVLFDPAZ-UHFFFAOYSA-N
SMILES: [Ce+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results