accessibility menu, dialog, popup

UserName

OReOOOAg

CAS RN 20654-56-2

IUPAC Name: silver(1+) rheniumoylolate
Synonyms: silver perrhenate
Molecular Weight (g/mol): 358.07
Molecular Formula: AgO4Re
InChi Key: JRISYOCHUNKJAV-UHFFFAOYSA-N
SMILES: [Ag+].[O-][Re](=O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results