accessibility menu, dialog, popup

UserName

CAS RN 20628-06-2

IUPAC Name: 1-(2-hydroxy-4,5-dimethoxyphenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-4,5-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: KEQHBVWVKYHDCS-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C(C=C1OC)C(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results