accessibility menu, dialog, popup

UserName

CAS RN 205877-26-5

CHN

CAS RN 205877-26-5

IUPAC Name: 4-ethynyl-N,N-diphenylaniline
Synonyms: 4-ethynyl-N,N-diphenylaniline
Molecular Weight (g/mol): 269.35
Molecular Formula: C20H15N
InChi Key: QDYFCLVSLUZDHB-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results