accessibility menu, dialog, popup

UserName

ClCH3NHCH3NH2H3C

CAS RN 2051-79-8

IUPAC Name: 4-amino-N,N-diethyl-3-methylanilinium chloride
Synonyms: 4-amino-N,N-diethyl-3-methylanilinium chloride
Molecular Weight (g/mol): 214.74
Molecular Formula: C11H19ClN2
InChi Key: MPLZNPZPPXERDA-UHFFFAOYSA-N
SMILES: [Cl-].CC[NH+](CC)C1=CC=C(N)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results