accessibility menu, dialog, popup

UserName

H3COSOOCH3

CAS RN 2051-05-0

IUPAC Name: dipentyl sulfite
Synonyms: dipentyl sulfite
Molecular Weight (g/mol): 222.34
Molecular Formula: C10H22O3S
InChi Key: QHKKGWHSXSIVAJ-UHFFFAOYSA-N
SMILES: CCCCCOS(=O)OCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results