accessibility menu, dialog, popup

UserName

OSOO

CAS RN 204905-77-1

IUPAC Name: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbaldehyde
Synonyms: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbaldehyde
Molecular Weight (g/mol): 170.18
Molecular Formula: C7H6O3S
InChi Key: GNVXYRDVJKJZTO-UHFFFAOYSA-N
SMILES: O=CC1=C2OCCOC2=CS1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)