accessibility menu, dialog, popup

UserName

OOOONaNaNaRu

CAS RN 20427-56-9

IUPAC Name: λ⁴-ruthenium(4+) trisodium tetraoxidandiide
Synonyms: ruthenium tetroxide
Molecular Weight (g/mol): 234.04
Molecular Formula: Na3O4Ru
InChi Key: QALKGQUGBXNZJX-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Na+].[Na+].[Na+].[Ru+4]
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results