accessibility menu, dialog, popup

UserName

CAS RN 204245-65-8

IUPAC Name: 3-(4-{[(tert-butoxy)carbonyl]amino}cyclohexyl)propanoic acid
Synonyms: 3-{4-[(tert-butoxycarbonyl)amino]cyclohexyl}propanoic acid
Molecular Weight (g/mol): 271.36
Molecular Formula: C14H25NO4
InChi Key: WCXYTBUDVLMXND-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CCC(O)=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results