accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOHOF

CAS RN 203866-14-2

IUPAC Name: 1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 233.24
Molecular Formula: C10H16FNO4
InChi Key: YGWZXQOYEBWUTH-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(F)CC1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results