accessibility menu, dialog, popup

UserName

CAS RN 20358-03-6

IUPAC Name: 5-bromo-1,3-benzothiazol-2-amine
Synonyms: 5-bromo-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 229.1
Molecular Formula: C7H5BrN2S
InChi Key: ZPUJTWBWSOOMRP-UHFFFAOYSA-N
SMILES: NC1=NC2=CC(Br)=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results