accessibility menu, dialog, popup

UserName

H3CCH3(Z)

CAS RN 20348-51-0

IUPAC Name: (2Z)-dec-2-ene
Synonyms: (2Z)-dec-2-ene
Molecular Weight (g/mol): 140.27
Molecular Formula: C10H20
InChi Key: YKNMBTZOEVIJCM-HYXAFXHYSA-N
SMILES: CCCCCCC\C=C/C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results