accessibility menu, dialog, popup

UserName

H3COOCH3OH3COOH(E/Z)

CAS RN 20329-98-0

IUPAC Name: 3-(3,4,5-trimethoxyphenyl)prop-2-enoic acid
Synonyms: TMCA O-methylsinapic acid
Molecular Weight (g/mol): 238.24
Molecular Formula: C12H14O5
InChi Key: YTFVRYKNXDADBI-UHFFFAOYSA-N
SMILES: COC1=CC(C=CC(O)=O)=CC(OC)=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results