accessibility menu, dialog, popup

UserName

H2NSS

CAS RN 20300-02-1

IUPAC Name: thiophene-2-carbothioamide
Synonyms: thiophene,2-thiocarboxamide
Molecular Weight (g/mol): 143.22
Molecular Formula: C5H5NS2
InChi Key: HDXYHAPUCGQOBX-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results