CAS RN 202982-65-8
IUPAC Name:
(2E)-3-(4-chloro-2-fluorophenyl)prop-2-enoate
Synonyms:
(2E)-3-(4-chloro-2-fluorophenyl)prop-2-enoate
Molecular Weight (g/mol):
199.59
Molecular Formula:
C9H5ClFO2
InChi Key:
FVLPOWWRHAOKMT-DUXPYHPUSA-M
SMILES:
[O-]C(=O)\C=C\C1=CC=C(Cl)C=C1F
1
–
1
of
1
results
1
4-Chloro-2-fluorocinnamic Acid 97.0+%, TCI America™
CAS: 202982-65-8 Molecular Formula: C9H5ClFO2 Molecular Weight (g/mol): 199.59 MDL Number: MFCD00143287 InChI Key: FVLPOWWRHAOKMT-DUXPYHPUSA-M Synonym: 4-chloro-2-fluorocinnamic acid,3-4-chloro-2-fluorophenyl acrylic acid,2e-3-4-chloro-2-fluorophenyl prop-2-enoic acid,4-chloro-2-fluorocinnamicacid,3-4-chloro-2-fluorophenyl prop-2-enoic acid,pubchem3431,e-3-4-chloro-2-fluorophenyl acrylic acid,rarechem bk hw 0062,timtec-bb sbb003542,trans-4-chloro-2-fluorocinnamic acid PubChem CID: 5941651 IUPAC Name: (2E)-3-(4-chloro-2-fluorophenyl)prop-2-enoate SMILES: [O-]C(=O)\C=C\C1=CC=C(Cl)C=C1F