CAS RN 202831-65-0
IUPAC Name:
4'-bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine
Synonyms:
4'-bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol):
400.32
Molecular Formula:
C24H18BrN
InChi Key:
NKCKVJVKWGWKRK-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Bromo-4'-(diphenylamino)biphenyl 93.0+%, TCI America™
CAS: 202831-65-0 Molecular Formula: C24H18BrN Molecular Weight (g/mol): 400.32 MDL Number: MFCD09038499 InChI Key: NKCKVJVKWGWKRK-UHFFFAOYSA-N Synonym: (4′C-Bromobiphenyl-4-yl)diphenylamine PubChem CID: 11315550 IUPAC Name: 4'-bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine SMILES: BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1