accessibility menu, dialog, popup

UserName

H3C(S)OHNH2CH3

CAS RN 2026-48-4

IUPAC Name: (2S)-2-amino-3-methylbutan-1-ol
Synonyms: L-valinol|2-amino-3-methyl-1-butanol|2-amino-3-methylbutan-1-ol
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: NWYYWIJOWOLJNR-RXMQYKEDSA-N
SMILES: CC(C)[C@H](N)CO
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)