accessibility menu, dialog, popup

UserName

CAS RN 2026-42-8

IUPAC Name: diethyl [(thiophen-2-yl)methyl]phosphonate
Synonyms: diethyl thiophen-2-ylmethylphosphonate
Molecular Weight (g/mol): 234.25
Molecular Formula: C9H15O3PS
InChi Key: GOJBUVQDOFHBLF-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC=CS1)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results