accessibility menu, dialog, popup

UserName

OOOOH3CH3CCH3CH3TaOH3COCH3

CAS RN 20219-33-4

IUPAC Name: tantalum(5+) (2Z)-4-oxopent-2-en-2-olate tetraethanolate
Synonyms: tantalum(5+) tetrakis(ethoxide) (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 460.3
Molecular Formula: C13H27O6Ta
InChi Key: LSALXRIVOAYYJW-GLUPTGNJSA-M
SMILES: [Ta+5].CC[O-].CC[O-].CC[O-].CC[O-].C\C([O-])=C\C(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)