accessibility menu, dialog, popup

UserName

CAS RN 202126-51-0

IUPAC Name: 5-hexyl-2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2-thione
Synonyms: 5-hexyl-5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2-thione
Molecular Weight (g/mol): 294.52
Molecular Formula: C10H14S5
InChi Key: RMWMTOWGAZGYGL-UHFFFAOYSA-N
SMILES: CCCCCCC1SC2=C(S1)SC(=S)S2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results