accessibility menu, dialog, popup

UserName

CAS RN 201481-37-0

H3C(S)O(S)HNOHOH3CCH3CH3OO

CAS RN 201481-37-0

IUPAC Name: (2S,3S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Synonyms: (2S,3S)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid|FMOC-ALLO-THR(TBU)-OH
Molecular Weight (g/mol): 397.47
Molecular Formula: C23H27NO5
InChi Key: LZOLWEQBVPVDPR-XOBRGWDASA-N
SMILES: C[C@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)