accessibility menu, dialog, popup

UserName

H3CCH3N(S)OHONHOOOOH3CH3CCH3

CAS RN 201003-48-7

IUPAC Name: (2S)-6-{[(tert-butoxy)carbonyl](propan-2-yl)amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-6-[(tert-butoxycarbonyl)(isopropyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid|N-FMOC-N'-BOC-N'-ISOPROPYL-L-LYSINE
Molecular Weight (g/mol): 510.63
Molecular Formula: C29H38N2O6
InChi Key: LUGFCMICCJNLBC-KSYWNVGFNA-N
SMILES: CC(C)N(CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)