accessibility menu, dialog, popup

UserName

CAS RN 2003-10-3

IUPAC Name: 2-bromo-1-[3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 267.05
Molecular Formula: C9H6BrF3O
InChi Key: TZIYNLSEBAYCBZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)CBr
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results