accessibility menu, dialog, popup

UserName

CAS RN 20010-99-5

IUPAC Name: 1-(pyrazin-2-yl)methanamine
Synonyms: 1-(pyrazin-2-yl)methanamine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: HQIBSDCOMQYSPF-UHFFFAOYSA-N
SMILES: NCC1=CN=CC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)