accessibility menu, dialog, popup

UserName

NHOOOHOOHH2O

CAS RN 199926-39-1

IUPAC Name: 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid hydrate
Synonyms: 4-oxo-1H-pyridine-2,6-dicarboxylic acid hydrate
Molecular Weight (g/mol): 201.13
Molecular Formula: C7H7NO6
InChi Key: SNGPHFVJWBKEDG-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=CC(=O)C=C(N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results