CAS RN 1999-00-4
IUPAC Name:
ethyl 3-(4-fluorophenyl)-3-oxopropanoate
Synonyms:
ethyl 3-(4-fluorophenyl)-3-oxopropanoate
Molecular Weight (g/mol):
210.2
Molecular Formula:
C11H11FO3
InChi Key:
SJUXLKYJKQBZLM-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(=O)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Ethyl (4-Fluorobenzoyl)acetate 98.0+%, TCI America™
CAS: 1999-00-4 Molecular Formula: C11H11FO3 Molecular Weight (g/mol): 210.204 MDL Number: MFCD03093631 InChI Key: SJUXLKYJKQBZLM-UHFFFAOYSA-N Synonym: ethyl 3-4-fluorophenyl-3-oxopropanoate,ethyl 4-fluorobenzoylacetate,ethyl 4-fluorobenzoyl acetate,4-fluorobenzoylacetic acid ethyl ester,ethyl p-fluorobenzoyl acetate,4-fluorobenzoyl acetic acid ethyl ester,ethyl 3-4-fluorophenyl-3-oxopropionate,ethyl 3-4'-fluorophenyl-3-oxopropanoate,ethyl 4'-fluorobenzoylacetate,3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester PubChem CID: 2758844 IUPAC Name: ethyl 3-(4-fluorophenyl)-3-oxopropanoate SMILES: CCOC(=O)CC(=O)C1=CC=C(C=C1)F