accessibility menu, dialog, popup

UserName

CH3NH2N

CAS RN 1990-90-5

IUPAC Name: 3-methylpyridin-4-amine
Synonyms: 3-methyl-4-aminopyridine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: VGJLGPCXUGIXRQ-UHFFFAOYSA-N
SMILES: CC1=CN=CC=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results