CAS RN 19883-77-3
IUPAC Name:
(2S)-2-amino-3-(3-fluorophenyl)propanoic acid
Synonyms:
3-fluorophenylalanine
Molecular Weight (g/mol):
183.18
Molecular Formula:
C9H10FNO2
InChi Key:
VWHRYODZTDMVSS-QMMMGPOBSA-N
SMILES:
N[C@@H](CC1=CC=CC(F)=C1)C(O)=O
1
–
1
of
1
results
1
3-Fluoro-L-phenylalanine 98.0+%, TCI America™
CAS: 19883-77-3 Molecular Formula: C9H10FNO2 Molecular Weight (g/mol): 183.182 MDL Number: MFCD00066450 InChI Key: VWHRYODZTDMVSS-QMMMGPOBSA-N Synonym: (S)-2-Amino-3-(3-fluorophenyl)propionic Acid, H-Phe(3-F)-OH PubChem CID: 716315 ChEBI: CHEBI:77957 IUPAC Name: (2S)-2-amino-3-(3-fluorophenyl)propanoic acid SMILES: C1=CC(=CC(=C1)F)CC(C(=O)O)N