accessibility menu, dialog, popup

UserName

OHONOH

CAS RN 19829-79-9

IUPAC Name: 8-hydroxyquinoline-7-carboxylic acid
Synonyms: 8-hydroxyquinoline-7-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: JYIAZVFJRYLCBH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=CC=NC2=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results