accessibility menu, dialog, popup

UserName

CAS RN 19780-84-8

IUPAC Name: hex-5-yn-3-ol
Synonyms: hex-5-yn-3-ol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: AJYGRAORQSCNED-UHFFFAOYNA-N
SMILES: CCC(O)CC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results