accessibility menu, dialog, popup

UserName

CAS RN 1978-37-6

IUPAC Name: 3-fluoro-N-methylaniline
Synonyms: 3-fluoro-N-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: FHYDHJXZZQCXOX-UHFFFAOYSA-N
SMILES: CNC1=CC=CC(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results