CAS RN 19658-77-6
IUPAC Name:
1-iodoisoquinoline
Synonyms:
1-iodoisoquinoline
Molecular Weight (g/mol):
255.06
Molecular Formula:
C9H6IN
InChi Key:
FDDBUIWRNBGXHB-UHFFFAOYSA-N
SMILES:
IC1=NC=CC2=CC=CC=C12
1
–
1
of
1
results
1
1-Iodoisoquinoline 98.0+%, TCI America™
CAS: 19658-77-6 Molecular Formula: C9H6IN Molecular Weight (g/mol): 255.06 MDL Number: MFCD00234494 InChI Key: FDDBUIWRNBGXHB-UHFFFAOYSA-N PubChem CID: 640964 IUPAC Name: 1-iodoisoquinoline SMILES: IC1=NC=CC2=CC=CC=C12