accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CBOOCH3CH3CH3CH3

CAS RN 196212-27-8

IUPAC Name: 4,4,5,5-tetramethyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 330.04
Molecular Formula: C18H28B2O4
InChi Key: LLQQCDJVSYEQQQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results