accessibility menu, dialog, popup

UserName

CAS RN 196-78-1

IUPAC Name: benzo[g]chrysene
Synonyms: benzo[g]chrysene
Molecular Weight (g/mol): 278.35
Molecular Formula: C22H14
InChi Key: JZOIZKBKSZMVRV-UHFFFAOYSA-N
SMILES: C1=CC=C2C(C=CC3=C2C2=CC=CC=C2C2=CC=CC=C32)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)