accessibility menu, dialog, popup

UserName

NH2HNOHOO

CAS RN 1956-25-8

IUPAC Name: 2-amino-3-[5-(benzyloxy)-1H-indol-3-yl]propanoic acid
Synonyms: 2-amino-3-[5-(benzyloxy)-1H-indol-3-yl]propanoic acid
Molecular Weight (g/mol): 310.35
Molecular Formula: C18H18N2O3
InChi Key: DFGNDJBYANKHIO-UHFFFAOYNA-N
SMILES: NC(CC1=CNC2=CC=C(OCC3=CC=CC=C3)C=C12)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results