accessibility menu, dialog, popup

UserName

SBr

CAS RN 19437-86-6

IUPAC Name: 2-(4-bromophenyl)-1-benzothiophene
Synonyms: 2-(4-bromophenyl)-1-benzothiophene
Molecular Weight (g/mol): 289.19
Molecular Formula: C14H9BrS
InChi Key: IJDKRWVYXSAROY-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results