accessibility menu, dialog, popup

UserName

CAS RN 19436-52-3

IUPAC Name: 2-acetamidopropanoic acid
Synonyms: 2-acetamidopropanoic acid
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: KTHDTJVBEPMMGL-UHFFFAOYNA-N
SMILES: CC(NC(C)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)