accessibility menu, dialog, popup

UserName

CAS RN 193357-52-7

IUPAC Name: 4-[(4-fluorophenyl)methyl]piperidine hydrochloride
Synonyms: 4-(4-fluorobenzyl)piperidine hydrochloride
Molecular Weight (g/mol): 229.72
Molecular Formula: C12H17ClFN
InChi Key: OJKWWANXBGLGQN-UHFFFAOYSA-N
SMILES: Cl.FC1=CC=C(CC2CCNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results