CAS RN 19315-93-6
IUPAC Name:
6-hydroxy-1,2-dihydroquinolin-2-one
Synonyms:
6-hydroxy-2-quinolone
Molecular Weight (g/mol):
161.16
Molecular Formula:
C9H7NO2
InChi Key:
AQLYZDRHNHZHIS-UHFFFAOYSA-N
SMILES:
OC1=CC=C2NC(=O)C=CC2=C1
1
–
1
of
1
results
1
2,6-Dihydroxyquinoline 98.0+%, TCI America™
CAS: 19315-93-6 Molecular Formula: C9H7NO2 Molecular Weight (g/mol): 161.16 MDL Number: MFCD00239446 InChI Key: AQLYZDRHNHZHIS-UHFFFAOYSA-N Synonym: 6-Hydroxy-2(1H)-quinolinone, 2,6-Quinolinediol PubChem CID: 177065 ChEBI: CHEBI:48996 IUPAC Name: 6-hydroxy-1H-quinolin-2-one SMILES: C1=CC2=C(C=CC(=O)N2)C=C1O